122769
Adipic acid monoethyl ester
97%
Synonym(s):
Mono-ethyl adipate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
HO2C(CH2)4CO2C2H5
CAS Number:
Molecular Weight:
174.19
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
97%
form
solid
refractive index
n20/D 1.439 (lit.)
bp
180 °C/18 mmHg (lit.)
mp
28-29 °C (lit.)
solubility
H2O: soluble 50 mg/mL, clear, colorless
density
0.98 g/mL at 25 °C (lit.)
SMILES string
CCOC(=O)CCCCC(O)=O
InChI
1S/C8H14O4/c1-2-12-8(11)6-4-3-5-7(9)10/h2-6H2,1H3,(H,9,10)
InChI key
UZNLHJCCGYKCIL-UHFFFAOYSA-N
Related Categories
Biochem/physiol Actions
Adipic acid monoethyl ester (AMME) suppresses the spore germination and mycelium development in phytopathogen Botrytis cinerea.AMME has antifungal properties.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
235.4 °F - closed cup
Flash Point(C)
113 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Preventive and post-infection control of Botrytis cinerea in tomato plants by hexanoic acid.
Leyva MO, et al.
Plant Pathology, 57(6), 1038-1046 (2008)
Youri M van Nuland et al.
Microbial cell factories, 16(1), 185-185 (2017-11-04)
Medium chain length (C6-C12) α,ω-dicarboxylic acids (DCAs) and corresponding esters are important building blocks for the polymer industry. For DCAs of 12 carbon atoms and longer, a sustainable process based on monooxygenase catalyzed ω-oxidation of fatty-acids has been realized. For
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service