Skip to Content
Merck
All Photos(1)

Key Documents

1235059

USP

Emodin

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Emodin, 1,3,8-Trihydroxy-6-methyl-9,10-anthracenedione, 1,3,8-Tri­hydroxy-6-methyl­anthra­quinone, 6-Methyl-1,3,8-tri­hydroxy­anthra­quinone, Archin, Emodol, Frangula-emodin, Frangulic acid, Rheum emodin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H10O5
CAS Number:
Molecular Weight:
270.24
Beilstein:
1888141
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

emodin

manufacturer/tradename

USP

application(s)

pharmaceutical

format

neat

storage temp.

2-8°C

SMILES string

Cc1cc(O)c2C(=O)c3c(O)cc(O)cc3C(=O)c2c1

InChI

1S/C15H10O5/c1-6-2-8-12(10(17)3-6)15(20)13-9(14(8)19)4-7(16)5-11(13)18/h2-5,16-18H,1H3

InChI key

RHMXXJGYXNZAPX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including MSDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

USP issued SDS can be found here.
Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service