Skip to Content
Merck
All Photos(1)

Key Documents

PHL82667

3′,4′,5,7-Tetrahydroxy-3,6,8-trimethoxyflavone

phyproof® Reference Substance

Synonym(s):

2-(3,4-Dihydroxyphenyl)-5,7-dihydroxy-3,6,8-trimethoxy-4H-1-benzopyran-4-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H16O9
CAS Number:
Molecular Weight:
376.31
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥95.0% (HPLC)

manufacturer/tradename

PhytoLab

format

neat

SMILES string

O1c2c(c(c(c(c2OC)O)OC)O)C(=O)C(=C1c3cc(c(cc3)O)O)OC

InChI

1S/C18H16O9/c1-24-16-11(21)10-12(22)17(25-2)14(7-4-5-8(19)9(20)6-7)27-15(10)18(26-3)13(16)23/h4-6,19-21,23H,1-3H3

InChI key

AOOSHVWNBJZOEH-UHFFFAOYSA-N

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service