58920-U
Rheodyne® Model 7000 Stream Switching Valve
Synonym(s):
Rheodyne 7000 Valve
All Photos(1)
About This Item
Recommended Products
feature
number of ports 6
positions 2
Quality Level
General description
Designed specifically for column selection, sample clean-up and enrichment, column programming, backflushing, and other stream switching operations. Six peripheral ports (no center port) are interconnected through a two-position rotor. Low volume flow passages (0.6 mm I.D.) minimize dead volume that can affect column switching analyses. The pressure limit is factory set at 5000 psi (350 kg/cm2), but can be adjusted to 7000 psi (490 kg/cm2).
The flat-face seal is easy to adjust.
The flat-face seal is easy to adjust.
Legal Information
Rheodyne is a registered trademark of IDEX Health & Science, LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)