Skip to Content
Merck
All Photos(9)

Key Documents

ZRB2377

Sigma-Aldrich

Anti-p-PKCβ (Ser660) Antibody, clone 2C11 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Anti-Ly-6C1 Antibody, clone 2H5 ZooMAb® Rabbit Monoclonal, EC:2.7.11.13, PKC-B, PKC-beta, Protein kinase C beta type

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

2C11, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 76.87 kDa
observed mol wt ~77 kDa

purified by

using Protein A

species reactivity

mouse, monkey

species reactivity (predicted by homology)

rat, rabbit, bovine, human

packaging

antibody small pack of 25

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
inhibition assay: suitable
western blot: suitable

isotype

IgG

epitope sequence

C-terminal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

Gene Information

human ... PRKCB(5579)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 2C11 is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects Protein Kinase C β (PKCβ) phosphorylated on serine 660.

Immunogen

KLH-conjugated linear peptide corresponding to 12 amino acids surrounding phosphoserine 660 from the C-terminal region of human Protein Kinase C β (PKCβ).

Application

Quality Control Testing

Evaluated by Western Blotting in lysate from NIH3T3 cells treated with phorbol 12-myristate 13-acetate (PMA).

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected PKC β phosphorylated on serine 660 in lysate from NIH3T3 cells treated with phorbol 12-myristate 13-acetate (PMA; 400 ng/mL; 20 min).

Tested Applications

Affinity Binding Assay: A representative lot of this antibody bound p-PKC β (Ser660) peptide with a KD of 9.8 x 10-7 in an affinity binding assay.

Peptide Inhibition Assay: Target band detection in lysate from NIH3T3 cells treated with phorbol 12-myristate 13-acetate (PMA; 400 ng/mL; 20 min) was prevented by preblocking of a representative lot with the immunogen phosphopeptide, but not the corresponding non-phosphopeptide.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected phospho-PKCβ (Ser660) in serum starved NIH3T3 cells treated with phorbol 12-myristate 13-acetate (PMA; 400 ng/ml) for 20 min.).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Protein kinase C beta type (UniProt: P05771; also known as EC:2.7.11.13, PKC-B, PKC-beta) is encoded by the PRKCB (also known as PKCB, PRKCB1) gene (Gene ID: 5579) in human. PKCβ is a calcium-activated, phospholipid- and diacylglycerol (DAG)-dependent serine/threonine kinase that is involved in multiple cellular processes, including the regulation of B-cell receptor, oxidative stress-induced apoptosis, insulin signaling, and endothelial cell proliferation. PKCβ is comprised of two splice variants, PKCβI and PKCβII, which are from the same gene and differ only in their last 50 amino acids. It binds three calcium ions per subunit and these ions are bound to the C2 domain (aa 158-275). Three specific sites: threonine 500 in the activation loop of the kinase domain; threonine 642 in the turn motif, and serine 661 in the hydrophobic region need to be phosphorylated for its full activation. Phosphorylation on threonine 500 renders it competent for autophosphorylation. Subsequent phosph

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Reconstitution

300 µg/mL after reconstitution at 25 µL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service