Skip to Content
Merck
All Photos(9)

Key Documents

ZRB1750

Sigma-Aldrich

Anti-LRBA Antibody, clone 3H22 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Lipopolysaccharide-responsive and beige-like anchor protein; Beige-like protein;CDC4-like protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

3H22, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 319.11 kDa
observed mol wt ~320 kDa

purified by

using Protein A

species reactivity

mouse, human

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

N-terminal half

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... LRBA(987)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 3H22 is a ZooMAb® Rabbit recombinant monoclonal antibody that specifically detects Lipopolysaccharide-responsive and beige-like anchor protein (LRBA). It targets and epitope within the N-terminal half.

Immunogen

His-tagged recombinant fragment corresponding to 244 amino acids from the N-terminal half of human Lipopolysaccharide-responsive and beige-like anchor protein (LRBA).

Application

Quality Control Testing

Evaluated by Western Blotting in U2OS cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected LRBA in U2OS cell lysate.

Tested applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected LRBA in NIH3T3 cell lysate.

Flow Cytometry Analysis: 0.1 μg from a representative lot detected LRBA in one million U2OS cells.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected LRBA in NIH3T3 cells.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Evaluated by Western Blotting in U2OS cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected LRBA in U2OS cell lysate.

Target description

Lipopolysaccharide-responsive and beige-like anchor protein (UniProt: P50851; also known as Beige-like protein, CDC4-like protein) is encoded by the LRBA (also known as BGL, CDC4L, LBA) gene (Gene ID: 987) in human. LRBA is a single-pass membrane protein that displays ubiquitous expression in normal tissues, but its expression is elevated in malignancy. In immune cells. It is found in all membrane compartments that are associated with receptor signaling, including the plasma membrane, Golgi complex, clathrin-coated pits, and trans-endocytic vesicles. Its expression is induced in B cells and macrophages by bacterial lipopolysaccharides (LPS). Two isoforms of LRBA have been reported that are produced by alternative splicing. LRBA contains 2 BEACH (Beige and Chediak-Higashi) domains and six WD repeats at its C-terminal. LRBA may be involved in coupling signal transduction and vesicle trafficking to enable polarized secretion and/or membrane deposition of immune effector molecules. It coordinates signaling of immune receptors to promote effector function and plays a crucial role in immune regulation. Mutations in LRBA gene lead to disorders of the immune system, including defective B-cell differentiation and decreased or absent antibody production, which results in early-childhood onset of recurrent infections. Mutations in LRBA gene are associated with defective B-cell differentiation and decreased or absent antibody production. This ZooMAbZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Park, MY et al. (2016). Nat. Sci. Rep. 6; 36568; Wang, J-W., et al. (2014). J. Allergy Clin. Immunol. 133(2; suppl); AB251).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

300 μg/mL after reconstitution at 25 μL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service