Skip to Content
Merck
All Photos(12)

Key Documents

ZRB1466

Sigma-Aldrich

Anti-Ryanodine Receptor 2 Antibody, clone 2K6 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

2K6, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 564.57 kDa

purified by

using Protein A

species reactivity

human

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunofluorescence: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG

epitope sequence

Internal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 2K6 is a ZooMAb® Rabbit recombinant monoclonal antibody that specifically detects human Ryanodine receptor 2. It targets an epitope within 20 amino acids from the internal region.

Immunogen

KLH-conjugated linear peptide corresponding to 20 amino acids from the internal region of human Ryanodine receptor 2.

Application

Quality Control Testing

Evaluated by Immunohistochemistry (Paraffin) in human heart tissue sections.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution of this antibody detected Ryanodine receptor 2 in human heart tissue sections.

Tested applications

Affinity Binding Assay: A representative lot of this antibody bound Ryanodine Receptor 2 with a KD of 9.2 x 10-9 in an affinity binding assay.

Immunofluorescence Analysis: A 1:100 dilution from a representative lot detected Ryanodine receptor 2 in human heart tissue sections.

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected Ryanodine receptor 2 in HeLa HiMark.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-Ryanodine Receptor 2, clone 2K6 ZooMAb®, Cat. No. ZRB1466, , is a recombinant Rabbit monoclonal antibody that targets Ryanodine receptor 2 and is tested for use in Affinity Binding Assay, Immunofluorescence, Immunohistochemistry, and Western Blotting.

Target description

Ryanodine receptor 2 (UniProt: Q92736; also known as RYR-2, RyR2, hRYR-2, Cardiac muscle ryanodine receptor, Cardiac muscle ryanodine receptor-calcium release channel, Type 2 ryanodine receptor) is encoded by the RYR2 gene (Gene ID: 6262) in human. Ryanodine receptors are large ion channels that are located in the sarcoplasmic/endoplasmic reticulum membrane and are responsible for the release of Ca2+. They form homotetramers with a mushroom-like shape, consisting of a large cytoplasmic head and transmembrane stalk. In mammals three isoforms of RYR are described (RYR 1-3) all of which are homoterameric proteins that are regulated by phosphorylation, redox modifications, and a variety of small proteins and ion. Ryanodine receptor 2 is a multi-pass membrane protein with two cytoplasmic domains (aa 1-4281 and 4871-4967), six transmembrane domains, and one pore-forming domain (aa 4820-4829). It is the predominant form in cardiac muscle and is also expressed in Purkinje cells of cerebellum and cerebral cortex. It plays a key role in triggering cardiac muscle contraction and aberrant channel activation can lead to cardiac arrhythmia. Channel activity is modulated by ryanodine that binds to the open Ca2+-release channel with high affinity. At low concentrations, ryanodine maintains the channel in an open conformation, but high concentrations of ryanodine inhibit channel activity. Channel activity is also modulated by phosphorylation. Phosphorylation at serine 2808 and serine 2814 is shown to increase the open probability of the calcium channel. Phosphorylation is increased in failing heart, leading to calcium leaks and increased cytoplasmic Ca2+ levels. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Lanner, JT., et al. (2010). Cold Spring Harb. Perspect. Biol. 2(11); a003996).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb® formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

300 μg/mL after reconstitution at 25 μL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service