Z768561
The Belly Dancer® orbital shaker
with tube mat and silicone mat, AC/DC input 230 V AC, Schuko plug
Synonym(s):
amplification, cell culture growth, gel staining, immunoprecipitation, membrane rinsing, northern hybridization, phage elution, southern hybridization, western blotting
About This Item
feature
0 ‑ 8 ° platform pitch control
manufacturer/tradename
IBI BDRLS0006
parameter
0-100 rpm speed
W × D × H
15 in. × 15 in. × 9 in.
platform W × L
12 in. × 12 in.
Looking for similar products? Visit Product Comparison Guide
General description
The smooth combination of vertical and horizontal orbital motion allows for more rapid exchange of reagents in applications such as staining and de-stainign gels and promotes lower background in blotting techniques. The unique 3-dimensional motion and wide range of speeds also provides superior growth in multiple cell culture and tissue culture applications. The easily adjustable speed and platform pitch angle controls provide a wide range of agitation conditions, from extremely gentle to vigorous.
The Belly Dancer is an exceptionally simple liquid agitation system, consisting of an orbiting platform flexibly anchored at four corners ao that the platform motion, when adjusted by the platform pitch control collar, is an extremely effective combination of rocking platform and orbital shaker. One-dimensional shakers do not have the versatility and range of application that result from the variable speed and adjustable platform pitch of the Belly Dancer.
To use this product in the UK please order Z768588 separately
Legal Information
required but not provided
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service