Z647128
BRAND® Transferpette® multichannel, electronic pipette
AC/DC input 115 V AC, US/Japan plug, 12 channel, volume 10-200 μL
About This Item
Recommended Products
feature
12-channel
CSA approved
accuracy± 0.8%
accuracy± 1.6 μL
electronic
graduations 0.2 μL
± 0.25% Coefficient of variation
± 0.5 μL Coefficient of variation
packaging
pack of 1 ea
manufacturer/tradename
BRAND 705474
tip size
200-300 μL
volume
10-200 μL
battery power
rechargeable
Looking for similar products? Visit Product Comparison Guide
General description
- Motor-driven to reduce the risk of RSI and imprecision/inaccuracy caused by operator variability
- Five modes: Pipetting, Reverse pipetting, Mixing, Dispensing, Gel-electrophoresis
- Easy and rapid calibration without tools
- The pipette shaft can be unscrewed and is entirely autoclavable (121 °C).
For pipettes up to 300 µl, includes: Transferpette®-8/-12 electronic, performance certificate, battery, power adapter, device stand, TipBox, refill pack, reagent reservoir, installation tool, silicone grease, and 1 set of sealing rings
For pipettes up to 1250 µl, includes: Transferpette®-8/-12 electronic, performance certificate, battery, power adapter, support frame, carousel stand mount, 2x TipBox, reagent reservoir, installation tool, grease and 1 set of sealing rings.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service