SRE0044
Trypsin inhibitor
lyophilized powder, suitable for mammalian cell culture
Synonym(s):
Trypsin inhibitor from Glycine max (soybean), SBTI
About This Item
Recommended Products
Product Name
Trypsin Inhibitor from Glycine max (soybean), Suitable for manufacturing of diagnostic kits and reagents, suitable for cell culture
biological source
Glycine max (soybean)
Quality Level
form
lyophilized powder
mol wt
14 kDa
technique(s)
cell culture | mammalian: suitable
solubility
0.067 M sodium phosphate buffer, pH 7.6: 1 mg/mL, clear, colorless to very faintly yellow
application(s)
diagnostic assay manufacturing
shipped in
wet ice
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
Application
Biochem/physiol Actions
Components
Unit Definition
Preparation Note
Solutions can retain activity when stored short-term at 2-8° C. Solutions are stable in frozen aliquots at -20°C.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Resp. Sens. 1 - Skin Sens. 1
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service