Skip to Content
Merck
All Photos(1)

Documents

SML0893

Sigma-Aldrich

M8-B hydrochloride

≥98% (HPLC)

Synonym(s):

N-(2-Aminoethyl)-N-(4-(benzyloxy)-3-methoxybenzyl)thiophene-2-carboxamide hydrochloride, N-(2-Aminoethyl)-N-[[3-methoxy-4-(phenylmethoxy)phenyl]methyl]-2-thiophenecarboxamide hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H24N2O3S · HCl
CAS Number:
Molecular Weight:
432.96
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: 2 mg/mL, clear (warmed)

storage temp.

2-8°C

InChI

1S/C22H24N2O3S.ClH/c1-26-20-14-18(9-10-19(20)27-16-17-6-3-2-4-7-17)15-24(12-11-23)22(25)21-8-5-13-28-21;/h2-10,13-14H,11-12,15-16,23H2,1H3;1H

InChI key

FMQJBHACRTZLME-UHFFFAOYSA-N

Application

M8-B hydrochloride has been used as a transient receptor potential melastatin-8 (TRPM8) channel antagonist to study its therapeutic efficiency on a mouse model of severe dry eye disease (DED) induced by the excision of extra-orbital lacrimal and Harderian glands.

Biochem/physiol Actions

M8-B is a selective and potent antagonist of the transient receptor potential melastatin-8 (TRPM8) channel. M8-B blocked cold-induced and icilin or methol-induced activation of rat, human, and murine TRPM8 channels with IC50 values in rat ranging from 7.8 nM to 64.3 nM, and it did not block other TRP channels (IC50 values > 20 mM). M8-B decreased deep body temperature in both mice and rats, showing that blockade of TRPM8 receptor might be used to induce therapeutic hypothermia, an effective treatment in cardiac arrest.

Features and Benefits

This compound is a featured product for Neuroscience research. Click here to discover more featured Neuroscience products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Darine Fakih et al.
International journal of molecular sciences, 21(22) (2020-11-25)
Dry eye disease (DED) is commonly associated with ocular surface inflammation and pain. In this study, we evaluated the effectiveness of repeated instillations of transient receptor potential melastatin 8 (TRPM8) ion channel antagonist M8-B on a mouse model of severe

Articles

DISCOVER Bioactive Small Molecules for Neuroscience

DISCOVER Bioactive Small Molecules for Neuroscience

DISCOVER Bioactive Small Molecules for Neuroscience

DISCOVER Bioactive Small Molecules for Neuroscience

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service