Key Documents
OH30087569
Ohaus® pH Electrode
STMICRO8
About This Item
UNSPSC Code:
41115604
packaging
pack of 1 ea
manufacturer/tradename
Ohaus® 30087569
pH measuring range
0.00-14 pH unit
General description
All electrodes include a temperature sensor, which powers automatic temperature compensation and ensures accurate measurements.
Application
pH, reference, oxidation-reduction potential (ORP), conductivity, dissolved oxygen (DO), temperature measurement
Features and Benefits
Can be used in conjunction with all Starter bench and portable meters
Legal Information
Ohaus is a registered trademark of Ohaus Corp.