MTCC3215SG
MTC™ Bio ClearSeal™ Graduated Screw Cap Microtubes
capacity 1.5 mL, sterile, cap, natural, self standing, pack of 1000 ea (20 x bags 50 ea)
Synonym(s):
centrifuge tube, screw-cap tube
All Photos(1)
About This Item
Recommended Products
material
clear
polypropylene
round
screw top cap
sterility
sterile
feature
cap natural
graduation
neck thread external
packaging
pack of 1000 ea (20 x bags 50 ea)
manufacturer/tradename
MTC™ Bio C3215-SG
parameter
22,000 × g speed
capacity
1.5 mL
diam. × L
12.8 mm × 47.6 mm
Related Categories
General description
ClearSeal™ screw-cap microtubes, 1.5mL , w/ O-ring & caps, sterile, printed graduations, self-standing, 20 resealable bags of 50 tubes, 1000/cs
Screw-cap microtubes include assembled caps with silicone O-rings that assure a leakproof seal from -86°C to +121°C and can be centrifuged at up to 22,000×g.
Features and Benefits
- Ultra-clear polypropylene
- Packaged in resealable bags of 50
- Leak-proof O-ring seal
- Sterile, RNase, DNase & Pyrogen free
- White, printed graduations
Legal Information
ClearSeal is a trademark of MTC Bio
MTC is a trademark of MTC Bio
Choose from one of the most recent versions:
Certificates of Analysis (COA)