Skip to Content
Merck
All Photos(2)

Key Documents

HPA006353

Sigma-Aldrich

Anti-SCUBE2 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Protein CEGP1, Anti-Signal peptide, CUB and EGF-like domain-containing protein 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50- 1:200

immunogen sequence

ESCGVGQGHAENQCVSCRAGTYYDGARERCILCPNGTFQNEEGQMTCEPCPRPGNSGALKTPEAWNMSECGGLCQPGEYSADGFAPCQLCALGTFQPEAGRTSC

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SCUBE2(57758)

General description

SCUBE2 (signal peptide, CUB domain, EGF-like 2) was originally recognized in endothelium and multiple nonendothelial primary cell types. In normal breasts, it is expressed in vascular endothelial and mammary ductal epithelial cells. It is a member of the evolutionary conserved small family called SCUBE. This family contains three members. SCUBe2 contains around five motifs namely, a leader peptide in its N-terminal, nine tandem EGF-like repeats, a large spacer region which is N-glycosylated, three repeated stretches of 6-cysteine residues, and a C-terminal with one CUB domain.

Immunogen

Signal peptide, CUB and EGF-like domain-containing protein 2 Precursor recombinant protein epitope signature tag (PrEST)

Application

Anti-SCUBE2 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

SCUBE2 (signal peptide, CUB domain, EGF-like 2) shows aberrant expression in breast cancer, and plays a role in tumorigenesis. It inhibits breast cancer cell proliferation, and its expression is linked to good prognosis. Thus, it might have potential as a marker for breast cancer. It inhibits breast tumor growth by acting as negative regulator of BMP (bone morphogenetic protein), and inhibiting the β-catenin pathway. It also prevents breast tumor invasion and migration by inhibiting epithelial-mesenchymal transition.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70280

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuh-Charn Lin et al.
Journal of cell science, 127(Pt 1), 85-100 (2013-11-12)
Signal peptide-CUB-EGF domain-containing protein 2 (SCUBE2) belongs to a secreted and membrane-associated multi-domain SCUBE protein family. We previously demonstrated that SCUBE2 is a novel breast-tumor suppressor and could be a useful prognostic marker. However, the role of SCUBE2 in breast-cancer
Yuh-Charn Lin et al.
The Journal of biological chemistry, 286(30), 27039-27047 (2011-06-10)
Signal peptide CUB (complement proteins C1r/C1s, Uegf, and Bmp 1)-EGF domain-containing protein 2 (SCUBE2) is a secreted, membrane-associated multidomain protein composed of five recognizable motifs: an NH(2)-terminal signal peptide sequence, nine copies of epidermal growth factor (EGF)-like repeats, a spacer
Chien-Jui Cheng et al.
Cancer research, 69(8), 3634-3641 (2009-04-17)
Signal peptide-CUB-epidermal growth factor-like domain-containing protein 2 (SCUBE2), originally identified from the endothelium and several nonendothelial primary cell types, was recently shown to be expressed in invasive breast carcinomas. However, the protein localization and biological significance of SCUBE2 in breast
Toshima Z Parris et al.
International journal of cancer, 134(7), 1617-1629 (2013-10-12)
The deregulation of key cellular pathways is fundamental for the survival and expansion of neoplastic cells, which in turn can have a detrimental effect on patient outcome. To develop effective individualized cancer therapies, we need to have a better understanding

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service