Key Documents
EPF740520015
Eppendorf® CryoCube® F740hi
capacity 740 L, touchscreen interface, cooling, 3, AC/DC input 120 - 208 V, 60 Hz, US 3-pin plug
Synonym(s):
CryoCube® F740hi, CryoCube® F740hi-ULT Freezer
About This Item
UNSPSC Code:
41103034
feature
cooling 3
touchscreen interface
capacity
740 L
General description
The Eppendorf CryoCube F740 series is one of the newest additions to the Eppendorf ultra-low temperature freezer family (also called ultra-low temperature freezer, -80°C). The CryoCube F740 series of ultra-low temperature freezers combines the traditionally high quality of our previous ultra-low temperature freezers with a higher capacity to safely store more of your samples while optimizing energy consumption. By incorporating the PhysioCare Concept into the development, the CryoCube F740 is lighter and more comfortable to use, supports ergonomics in the workplace and enables an optimal workflow in the laboratory.
Legal Information
CryoCube is a registered trademark of Eppendorf AG
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)