EP022491156
Eppendorf® epT.I.P.S. Biopur®,singles
volume 50-1,000 μL, L 71 mm, blue
Synonym(s):
pipette tips
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121600
volume:
50-1,000 μL
sterility:
sterile
feature:
DNase free
RNase free
endotoxin free
RNase free
endotoxin free
packaging:
pack of 100 ea
manufacturer/tradename:
Eppendorf® 022491156
material
blue
polypropylene
sterility
sterile
feature
DNase free
RNase free
endotoxin free
packaging
pack of 100 ea
manufacturer/tradename
Eppendorf® 022491156
L
71 mm
volume
50-1,000 μL
Looking for similar products? Visit Product Comparison Guide
General description
epT.I.P.S.® Singles, Biopur®, 50 – 1,000 µL, 71 mm, blue, colorless tips, 100 tips, individually packed. Replaces order no. 0030010051.
- Eppendorf Biopur pipette tip, individually wrapped
- Ensured PCR clean, sterile, pyrogen-, ATP- and DNA-free
- For use in pharmaceutical and food industry, molecular biology and cell technology
- Batch number and expiration date on each blister package
- Each production lot inspected continuously by an independent laboratory
- Lot-specific certificates available at www.eppendorf.com/certificates
Features and Benefits
- Eppendorf Biopur pipette tip, individually wrapped
- Ensured PCR clean, sterile, pyrogen-, ATP- and DNA-free
- For use in pharmaceutical and food industry, molecular biology and cell technology
- Batch number and expiration date on each blister package
- Each production lot inspected continuously by an independent laboratory
- Lot-specific certificates available at www.eppendorf.com/certificates
Legal Information
Biopur is a registered trademark of Eppendorf AG
Eppendorf is a registered trademark of Eppendorf AG
epT.I.P.S. is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service