CLS7016560
Corning® Soda Lime Silica glass petri dish
dish diam. 60 mm × 15 mm
Synonym(s):
Filter crucible
All Photos(1)
About This Item
UNSPSC Code:
41121812
NACRES:
NB.43
Recommended Products
material
round
soda-lime glass dishes with lids
sterility
non-sterile
feature
lid
reusable
packaging
case of 24
manufacturer/tradename
Corning 70165-60
dish diam.
60 mm × 15 mm
Related Categories
General description
The thermal expansion of Soda Lime Glass is 93.5 x 10-7 cm/cm/°C. The normal service temperature is 110°C and the extreme service temperature is 460°C. These dishes are not designed for rapid thermal shock. The Corning® brand Petri Dishes should be viewed as a low cost alternative to Pyrex® brand Petri Dishes (Catalog CLS3160) which are made from low expansion (32.5 x 10-7 cm/cm/°C) borosilicate glass.
Legal Information
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)