Skip to Content
Merck
All Photos(1)

Documents

NCI-H322 Cell Line human

95111734, human lung (cervical node metastasis), Not specified

Synonym(s):

H322 Cells, NCI-322 Cells, NCI-H322T Cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

NCI-H322 Cell Line human, 95111734

biological source

human lung (cervical node metastasis)

description

Human Caucasian bronchioalveolar carcinoma

growth mode

Adherent

karyotype

Not specified

morphology

Not specified

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

metastasis

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human Caucasian bronchioalveolar carcinoma

Cell Line Description

NCI-H322 was derived in 1981 from a primary bronchioalveolar carcinoma of the lung from a 52 year old male taken prior to treatment. Ultrastructural studies of this non-small cell carcinoma cell line demonstrated the presence of cytoplasmic structures characteristic of Clara cells. NCI-H322 do not express UDP-glucuronosyltransferase, but are reported to express glutathione-S-transferase and phenol sulfotransferase. Expression of protein and RNA of SP-A, the major surfactant-associated protein was detected. The cells produce tumours in athymic mice.

Application

NCI-H322 has been used to study the preclinical antitumor efficacy of BAY 1129980, which is an anti-C4.4A (Ly6/PLAUR domain-containing protein 3 precursor-LYPD3) antibody–drug conjugate for the treatment of non–small cell lung cancer.
Test system for evaluating cytotoxicity and genotoxicity of chemicals to human lung

Culture Medium

RPMI 1640 + 2mM Glutamine + 5-10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 1-3x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service