Skip to Content
Merck
All Photos(1)

Key Documents

XC

88120601, rat muscle, Epithelial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

Product Name

XC, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA., 88120601

biological source

rat muscle

packaging

tube of 5 μg 88120601-DNA-5UG
pkg of vial of cells 88120601-1VL

growth mode

Adherent

karyotype

2n = 42

morphology

Epithelial

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Rat Wistar sarcoma

Cell Line Description

Original tumour induced in outbred Wistar rats by intramuscular injection of Rous sarcoma virus. The cells have virus genome but do not normally release infectious virus. The XC plaque assay is an indirect method of detection for MuLV in cultured cells.

Application

Plaque assay and virus studies: Rous sarcoma virus, MuLV

Culture Medium

EMEM (HBSS) + 2mM Glutamine + 1% Non Essential Amino Acids (NEAA) + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:2 to 1:6 i.e. seeding at 2-5x10,000 cells/cm2 using 0.25% trypsin; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service