Skip to Content
Merck
All Photos(1)

Key Documents

A673

85111504, human muscle, Epithelial-like

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
biological source:
human muscle
growth mode:
Adherent
karyotype:
Near Diploid
morphology:
Epithelial-like
products:
Transforming growth factor (Tramnsforming Growth Factor (TGF)) - ß
receptors:
Not specified
relevant disease(s):
cancer

Product Name

A673, 85111504

biological source

human muscle

growth mode

Adherent

karyotype

Near Diploid

morphology

Epithelial-like

products

Transforming growth factor (Tramnsforming Growth Factor (TGF)) - ß

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human Ewing sarcoma

Cell Line Description

Derived from a 15 year old female patient. The cells form colonies on soft agar and tumours in antithymocyte serum treated mice.

The cell line A673 was originally described in 1973 as derived from a rhabdomyosarcoma, however, more recent analysis of the cells using advanced diagnostic techniques determined the cells to have characteristics of Ewing sarcoma rather than rhabdomyosarcoma. See citations for more information about this tumour status.

Application

Tumourigenicity studies

DNA Profile

STR-PCR Data: Amelogenin: X
CSF1PO: 11,12
D13S317: 8,13
D16S539: 11
D5S818: 11,12
D7S820: 10,12
THO1: 9.3
TPOX: 8
vWA: 15,18

Culture Medium

DMEM + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:10, i.e., seeding at 1-3x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37 °C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service