COV434
NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.
Synonym(s):
COV-434 Cells
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106514
Recommended Products
biological source
human ovary
packaging
tube of 5 μg 07071909-DNA-5UG
pkg of vial of cells 07071909-1VL
growth mode
Adherent
karyotype
Modal chromosome no. 47 (46 - 47)
morphology
Polygonal & fusiform
products
Not specified
receptors
Not specified
technique(s)
cell culture | mammalian: suitable
relevant disease(s)
cancer
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Human ovarian granulosa tumour
Cell Line Description
Human ovarian granulosa tumour cell line established from a solid primary tumour.
Application
COV434 has been used to study:
- the effect of 17β-estradiol on granulosa cell tumor growth, migration and invasion
- if mutant p53 (inactivatible R248Q hotspot mutation) works as a cancer-specific drug target
- the expression of TLR (toll-like receptor) and its function in the cell line
Culture Medium
DMEM + 2mM Glutamine + 10% Foetal Bovine Serum (FBS)
Subculture Routine
Split confluent cultures 1:2 to 1:5 i.e. seeding at 2-4x10,000 cells/cm2 using trypsin/EDTA; 5% CO2; 37°C.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service