Skip to Content
Merck
All Photos(1)

Key Documents

B0555

Sigma-Aldrich

Anti-Vascular Endothelial Growth Factor Receptor-2 (KDR)−Biotin Conjugate antibody, Mouse monoclonal

clone 260.4, KDR-2, purified from hybridoma cell culture

Synonym(s):

Monoclonal Anti-Vascular Endothelial Growth Factor Receptor-2 antibody produced in mouse, Anti-KDR, Anti-VEGFR-2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
51111800
NACRES:
NA.41
conjugate:
biotin conjugate
application:
WB
clone:
260.4, KDR-2, monoclonal
species reactivity:
human
citations:
9
technique(s):
western blot: 1:1,000 using insect cell culture supernatant expressing human extracellular recombinant VEGF R

biological source

mouse

Quality Level

conjugate

biotin conjugate

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

260.4, KDR-2, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

western blot: 1:1,000 using insect cell culture supernatant expressing human extracellular recombinant VEGF R

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... KDR(3791)

Looking for similar products? Visit Product Comparison Guide

General description

Biotin Conjugated Monoclonal Anti-VEGF Receptor-2 (KDR) (mouse IgG1 isotype) is derived from the KDR-2 hybridoma produced by the fusion of mouse myeloma cells and splenocytes from a BALB/c mouse immunized with a purified recombinant human extracellular VEGF receptor-2 (KDR). Vascular endothelial growth factor (VEGF) is also called vasculotropin (VAS), vascular permeability factor (VPF) and kinase-insert domain containing receptor (KDR8). It is a member of a family of endothelial cell mitogens and angiogenic factors. VEGF is a homodimeric heparin-binding glycoprotein. The KDR gene has been mapped to human chromosome 4q11-q12. The homologous gene for the human KDR gene is mouse Flk1 (fetal liver kinase 1) and rat TKrc.

Specificity

Monoclonal Anti-Vascular Endothelial Growth Factor Receptor-2 (KDR)-Biotin Conjugate antibody is purified from ascites fluid, and conjugated withSulfo-NHS-LC-Biotin. It is specific for human VEGF receptor2. It does not cross react with Flt1, Flt4 or platelet-derived growth factor receptor β (PDGF-R1).
Reacts specifically with human VEGF receptor-2. This clone has also been termed 260.4. It does not recognize Flt1 (VEGFR-1), Flt4 (VEGFR-3), or platelet-derived growth factor receptor (PDGF-R).

Immunogen

purified recombinant human extracellular VEGF receptor-2 (KDR).

Application

Monoclonal Anti-Vascular Endothelial Growth Factor Receptor-2 (KDR)-Biotin Conjugate antibody produced in mouse has also been used in :
  • enzyme linked immunosorbent assay (ELISA)
  • immunoblotting
  • flow cytometry
  • immunohistochemistry
  • immunohistochemistry

Biochem/physiol Actions

Vascular endothelial growth factors receptors (VEGF Rs) are receptor tyrosine kinases that bind to VEGF that plays important role in embryonic vasculogenesis, angiogenesis and homeostasis. VEGF R2 has the ability to form heterodimers with VEGF R1 and VEGF R3. The binding of VEGF to VEGF R2 results in activation of tyrosine amino acids of the cytoplasmic tail of the receptor. This interaction results in the activation of MAPK, PI3K, PKC, FAK and Src kinase pathways. The biological roles of VEGF R2 include in epithelial cell growth, proliferation, survival, migration and vascular permeability and angiogenesis. Due to its role in promoting migration and angiogenesis, VEGF R2 has been implicated in breast and prostate cancer.

Target description

The KDR receptor (VEGFRa), defining marker of hematopoietic stem cells (HSCs), distinguishes pluripotent HSCs from lineage-committed progenitor HSCs.

Physical form

Solution in phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin and 15 mM sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Böldicke et al.
Stem cells (Dayton, Ohio), 19(1), 24-36 (2001-02-24)
Five specific single-chain antibodies recognizing the human vascular endothelial growth factor receptor-2 (VEGFR-2/KDR) were selected from a V-gene phage display library constructed from mice immunized with the extracellular domain of VEGFR-2 (Ig-like domain 1-7). All five scFv antibodies (A2, A7
Shanchun Guo et al.
Biochimica et biophysica acta, 1806(1), 108-121 (2010-05-14)
Investigations over the last decade have established the essential role of growth factors and their receptors during angiogenesis and carcinogenesis. The vascular endothelial growth factor receptor (VEGFR) family in mammals contains three members, VEGFR-1 (Flt-1), VEGFR-2 (KDR/Flk-1) and VEGFR-3 (Flt-4)
P Berger et al.
Swiss medical weekly, 141, w13318-w13318 (2011-12-20)
Vascular endothelial growth factors (VEGFs) regulate blood and lymph vessel formation through activation of the type V receptor tyrosine kinases VEGFR-1, -2 and -3. In addition, VEGFs interact with co-receptors such as neuropilins, integrins, semaphorins or heparansulfate glycosaminoglycans. Ligand binding
Hyperoxia reduces bone marrow, circulating, and lung endothelial progenitor cells in the developing lung: implications for the pathogenesis of bronchopulmonary dysplasia
Balasubramaniam V, et al.
American Journal of Physiology. Lung Cellular and Molecular Physiology, 292(5), L1073-L1084 (2007)
Role of vascular endothelial growth factor in regulation of physiological angiogenesis
Ferrara N
American Journal of Physiology. Cell Physiology, 280(6), C1358-C1366 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service