Skip to Content
Merck
All Photos(2)

Key Documents

AV44694

Sigma-Aldrich

Anti-MPG antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-AAG, Anti-APNG, Anti-CRA36.1, Anti-MDG, Anti-Mid1, Anti-N-Methylpurine-DNA glycosylase, Anti-PIG11, Anti-PIG16

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

32 kDa

species reactivity

rat, mouse, human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MPG(4350)

Immunogen

Synthetic peptide directed towards the C terminal region of human MPG

Application

Anti-MPG antibody produced in rabbit is suitable for western blotting at a concentration of 0.5μg/ml.

Biochem/physiol Actions

N-methylpurine-DNA glycosylase (MPG) is an enzyme that repairs the genome-wide damage caused by alkylating agents. MPG repairs the hypoxanthine that is formed by the deamination of adenine and 1,N6-ethenoadenine formed as a result of interaction of lipid peroxidation-derived aldehydes and hydroxyalkenals with DNA. A functional linkage between MPG and p53; MPG acting as the selective regulator of p53-mediated cell cycle arrest.

Sequence

Synthetic peptide located within the following region: LEPSEPAVVAAARVGVGHAGEWARKPLRFYVRGSPWVSVVDRVAEQDTQA

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Robyn L Maher et al.
DNA repair, 6(1), 71-81 (2006-10-05)
The base excision repair pathway removes damaged DNA bases and resynthesizes DNA to replace the damage. Human alkyladenine DNA glycosylase (AAG) is one of several damage-specific DNA glycosylases that recognizes and excises damaged DNA bases. AAG removes primarily damaged adenine
Yael Leitner-Dagan et al.
Journal of the National Cancer Institute, 104(22), 1765-1769 (2012-10-30)
Only a minority of smokers develop lung cancer, possibly due to genetic predisposition, including DNA repair deficiencies. To examine whether inter-individual variations in DNA repair activity of N-methylpurine DNA glycosylase (MPG) are associated with lung cancer, we conducted a blinded
Chun-Yue I Lee et al.
Biochemistry, 48(9), 1850-1861 (2009-02-18)
The human 3-methyladenine DNA glycosylase (AAG) recognizes and excises a broad range of purines damaged by alkylation and oxidative damage, including 3-methyladenine, 7-methylguanine, hypoxanthine (Hx), and 1,N(6)-ethenoadenine (epsilonA). The crystal structures of AAG bound to epsilonA have provided insights into
Shanshan Song et al.
Cell research, 22(8), 1285-1303 (2012-07-18)
Alkylating agents induce genome-wide base damage, which is repaired mainly by N-methylpurine DNA glycosylase (MPG). An elevated expression of MPG in certain types of tumor cells confers higher sensitivity to alkylation agents because MPG-induced apurinic/apyrimidic (AP) sites trigger more strand

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service