If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf
Recommended Products
grade
certified reference material
Quality Level
manufacturer/tradename
NIST®
format
matrix material
SMILES string
[Si](=O)=O.[Al](=O)O[Al]=O
InChI
1S/2Al.O2Si.3O/c;;1-3-2;;;
InChI key
HNPSIPDUKPIQMN-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
SRM 8850_cert
SRM 8850_SDS
Application
Features and Benefits
- Available with the certificate documenting NIST-certified reference mass fraction values for major components of Zeolite Y.
- Information mass fraction values of elements are provided.
- The NIST certificate is provided with expiration certificate, storage, handling, use, and maintenance instructions.
Other Notes
Sodium (Na), Aluminium (Al), Silicon (Si), Iron (Fe), Cerium (Ce), Europium (Eu), Hafnium (Hf), Lanthanum (La), Rubidium (Rb), Scandium (Sc), Samarium (Sm), Thorium (Th), Zinc (Zn), Cobalt (Co), Chromium (Cr), Cesium (Cs)
See certificate for values and more details at nist.gov/SRM.
Legal Information
related product
Storage Class Code
13 - Non Combustible Solids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
-
How can I determine the shelf life / expiration / retest date of this product?
1 answer-
Helpful?
-
-
How is shipping temperature determined? And how is it related to the product storage temperature?
1 answer-
Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf
Helpful?
-
-
What will be the Silicaon Aluminium ratio of this product and what are the different types of Zeolite USY you can provide us ?
1 answer-
This product has 22.52% Silicon and 8.49% mass fraction. Please refer to the COA found here: https://tsapps.nist.gov/srmext/certificates/8850.pdf. Here are the other Zeolite products available in the catalog: https://www.sigmaaldrich.com/US/en/substance/123451318021.
Helpful?
-
-
Hi, Can I get a full data sheet for this product please? Regards, Nazim
1 answer-
Please see the available supplier documentation below:
https://tsapps.nist.gov/srmext/certificates/8850.pdf
https://tsapps.nist.gov/srmext/msds/8850-MSDS.pdfHelpful?
-
-
What is the density of this catalyst ?
1 answer-
The SDS literature value is relative density 2.2 at 20 °C (68 °F). More extensive information can be found in the SRM certification: https://www-s.nist.gov/srmors/certificates/8850.pdf.
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service