NIST705A
Polystyrene (narrow molecular weight distribution)
NIST® SRM® 705a
Synonym(s):
Polystyrene, PS
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
grade
certified reference material
Quality Level
form
solid
packaging
pkg of 5 g
manufacturer/tradename
NIST®
application(s)
chemicals and industrial polymers
Looking for similar products? Visit Product Comparison Guide
General description
Application
The Polystyrene SRM is intended to calibrate instruments used to determine molecular weight in polymer science and technology to check dynamic thermal analytical instruments.
Other Notes
Example analytes are listed below as a reference. Please download a current certificate at nist.gov/SRM for current analytes and certified values.
Polystyrene
See certificate for values and more details at nist.gov/SRM.
Polystyrene
See certificate for values and more details at nist.gov/SRM.
Legal Information
NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service