All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C7H5FO2
CAS Number:
Molecular Weight:
140.11
UNSPSC Code:
41116107
NACRES:
NA.24
Recommended Products
grade
certified reference material
packaging
pkg of 2 g
manufacturer/tradename
NIST®
format
neat
storage temp.
15-25°C
InChI
1S/C7H5FO2/c8-6-3-1-5(2-4-6)7(9)10/h1-4H,(H,9,10)
InChI key
BBYDXOIZLAWGSL-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Other Notes
Certified for the analytes listed below.
Fluorine (F)
See certificate for values and more details at nist.gov/SRM.
Fluorine (F)
See certificate for values and more details at nist.gov/SRM.
Legal Information
NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology
related product
Product No.
Description
Pricing
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service