Skip to Content
Merck
All Photos(1)

Key Documents

939021

Sigma-Aldrich

Sodium carbonate ChemBeads

Synonym(s):

Soda ash ChemBeads, Calcined soda ChemBeads, Carbonic acid disodium salt ChemBeads

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Na2CO3
CAS Number:
Molecular Weight:
105.99
MDL number:
UNSPSC Code:
12352100
Pricing and availability is not currently available.

description

Inorganic salt

Quality Level

form

solid

composition

, 14-16 wt. % (loading of base)

reaction suitability

core: sodium

SMILES string

O=COO[Na].[Na]

InChI

1S/CH2O3.2Na/c2-1(3)4;;/h(H2,2,3,4);;/q;2*+1/p-2

InChI key

CDBYLPFSWZWCQE-UHFFFAOYSA-L

General description

Sodium carbonate (soda ash) is an inorganic chemical compound. It is used in the synthesis of chemicals, detergent formulations, flue gas desulfurization, and paper and pulp production.

Application

Sodium carbonate can be used: As a catalyst in the aldol condensation of aldehydes with ketones in the synthesis of chalcone and azachalcone. As a pH stabilizer agent in the synthesis of FeO-NPs (iron oxide nanoparticles) in the presence of cymbopogon citratus as a reducing agent. As a base in the Pd-catalyzed Suzuki coupling reactions. As a base in the transition metal free Suzuki coupling reaction of aryl halides and arylboronic acids to form biaryls using water as a solvent and TBAB (tetrabutylammonium bromide) as a phase-transfer catalyst. For general uses, product is also available in powdered form (223530)

Features and Benefits

ChemBeads are chemical coated glass beads. ChemBeads offer improved flowability and chemical uniformity perfect for automated solid dispensing and high-throughput experimentation. The method of creating ChemBeads uses no other chemicals or surfactants allowing the user to accurately dispense sub-milligram amounts of chemical.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ji Zhang et al.
Nature communications, 6, 6615-6615 (2015-03-20)
Commercial lead-based piezoelectric materials raised worldwide environmental concerns in the past decade. Bi₀.₅Na₀.₅TiO₃-based solid solution is among the most promising lead-free piezoelectric candidates; however, depolarization of these solid solutions is a longstanding obstacle for their practical applications. Here we use
Ana L Aguirre et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 27(51), 12981-12986 (2021-07-08)
High-throughput experimentation (HTE) methods are central to modern medicinal chemistry. While many HTE approaches to C-N and Csp2 -Csp2 bonds are available, options for Csp2 -Csp3 bonds are limited. We report here how the adaptation of nickel-catalyzed cross-electrophile coupling of
Noah P Tu et al.
Angewandte Chemie (International ed. in English), 58(24), 7987-7991 (2019-03-21)
Technologies that enable rapid screening of diverse reaction conditions are of critical importance to methodology development and reaction optimization, especially when molecules of high complexity and scarcity are involved. The lack of a general solid dispensing method for chemical reagents
Hyojung Cha et al.
ACS applied materials & interfaces, 6(18), 15774-15782 (2014-08-26)
The molecular packing structures of two conjugated polymers based on alkoxy naphthalene, one with cyano-substituents and one without, have been investigated to determine the effects of electron-withdrawing cyano-groups on the performance of bulk-heterojunction solar cells. The substituted cyano-groups facilitate the
Chenghong Wang et al.
Scientific reports, 5, 16613-16613 (2015-11-13)
In this study, water stable zirconium metal-organic framework (UiO-66) has been synthesized and for the first time applied as an adsorbent to remove aquatic arsenic contamination. The as-synthesized UiO-66 adsorbent functions excellently across a broad pH range of 1 to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service