Skip to Content
Merck
All Photos(1)

Documents

722545

Sigma-Aldrich

3-Bromopyruvic acid-3-13C

99 atom % 13C, 97% (CP)

Synonym(s):

3-Bromo-2-oxopropionic acid-3-13C

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Br13CH2COCOOH
Molecular Weight:
167.95
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:

isotopic purity

99 atom % 13C

Quality Level

Assay

97% (CP)

form

powder

mass shift

M+1

storage temp.

−20°C

SMILES string

OC(=O)C(=O)[13CH2]Br

InChI

1S/C3H3BrO3/c4-1-2(5)3(6)7/h1H2,(H,6,7)/i1+1

InChI key

PRRZDZJYSJLDBS-OUBTZVSYSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service