Skip to Content
Merck
All Photos(2)

Key Documents

40820

Supelco

Chebulinic acid

analytical standard

Synonym(s):

Eutannin, NSC 69862

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C41H32O27
CAS Number:
Molecular Weight:
956.68
MDL number:
UNSPSC Code:
85151701
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥90.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

format

neat

storage temp.

2-8°C

SMILES string

O1[C@@H]2[C@H](O[C@H]([C@H](OC(=O)c6c7c(c(c(c6)O)O)OC(=O)[C@H]([C@H]7[C@@H](C1=O)CC(=O)O)O)[C@H]2OC(=O)c5cc(c(c(c5)O)O)O)OC(=O)c4cc(c(c(c4)O)O)O)COC(=O)c3cc(c(c(c3)O)O)O

InChI

1S/C41H32O27/c42-15-1-10(2-16(43)26(15)51)35(56)62-9-22-31-33(66-36(57)11-3-17(44)27(52)18(45)4-11)34(41(63-22)68-37(58)12-5-19(46)28(53)20(47)6-12)67-38(59)13-7-21(48)29(54)32-25(13)24(30(55)40(61)65-32)14(8-23(49)50)39(60)64-31/h1-7,14,22,24,30-31,33-34,41-48,51-55H,8-9H2,(H,49,50)/t14-,22+,24-,30-,31+,33-,34+,41-/m0/s1

InChI key

YGVHOSGNOYKRIH-FJPMMHPYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service