Skip to Content
Merck
All Photos(1)

Key Documents

28-0240

Sigma-Aldrich

Samson′s solution

Synonym(s):

Samson’s reagent solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12000000
form:
liquid

form

liquid

availability

available only in Japan

storage temp.

15-25°C

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Muta. 2 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service