Skip to Content
Merck
All Photos(1)

Documents

13-2620

Sigma-Aldrich

Hypophosphorous acid solution

SAJ first grade, 30.0-32.0% in H2O

Synonym(s):

Phosphinic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H3PO2
CAS Number:
Molecular Weight:
66.00
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:

grade

SAJ first grade

form

liquid

availability

available only in Japan

concentration

30.0-32.0% in H2O

SMILES string

O[PH2]=O

InChI

1S/H3O2P/c1-3-2/h3H2,(H,1,2)

InChI key

ACVYVLVWPXVTIT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Met. Corr. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anne M J Verstegen et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 34(21), 7266-7280 (2014-05-23)
Cyclin-dependent kinase-5 (Cdk5) was reported to downscale neurotransmission by sequestering synaptic vesicles (SVs) in the release-reluctant resting pool, but the molecular targets mediating this activity remain unknown. Synapsin I (SynI), a major SV phosphoprotein involved in the regulation of SV
Shuhei Kobuchi et al.
Clinical and experimental pharmacology & physiology, 42(3), 278-286 (2014-12-09)
Excitation of the renal sympathetic nervous system is important for the development of ischaemic acute kidney injury (AKI) in rats. We reported that intravenous treatment with GABA has preventive effects against ischaemia/reperfusion (I/R)-induced renal dysfunction with histological damage in rats;
Yuqing Liu et al.
Scientific reports, 5, 17045-17045 (2015-11-21)
Although great attention has been paid to wearable electronic devices in recent years, flexible lightweight batteries or supercapacitors with high performance are still not readily available due to the limitations of the flexible electrode inventory. In this work, highly flexible
Chenchen Li et al.
The Journal of physiology, 592(19), 4329-4351 (2014-08-12)
Competing mechanisms of long-term potentiation (LTP) and long-term depression (LTD) in principal neurons of the basolateral amygdala (BLA) are thought to underlie the acquisition and consolidation of fear memories, and their subsequent extinction. However, no study to date has examined
Puneet Sodhi et al.
The Journal of physiology, 592(19), 4201-4220 (2014-07-20)
Adenosine is an established neuromodulator in the mammalian retina, with A1 adenosine receptors being especially prevalent in the innermost ganglion cell layer. Activation of A1 receptors causes inhibition of adenylate cyclase, decreases in intracellular cyclic AMP (cAMP) levels and inhibition

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service