Skip to Content
Merck
All Photos(1)

Key Documents

05-1251

Sigma-Aldrich

(1S)-(+)-10-Camphorsulfonic acid ammonium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16O4S · H3N
Molecular Weight:
249.33
UNSPSC Code:
12352106
grade:
SAJ special grade
form:
solid

grade

SAJ special grade

form

solid

availability

available only in Japan

Suitability

circular dichroism determination

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service