Skip to Content
Merck
All Photos(5)

Key Documents

SCC487

Sigma-Aldrich

CNS-1 Rat Glioma Cell Line

Rat

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
NACRES:
NA.81
biological source:
rat
growth mode:
N/A
manufacturer/tradename:
Millipore

Product Name

CNS-1 Rat Glioma Cell Line,

biological source

rat

Quality Level

packaging

vial of ≥1X10⁶ cells/vial mg

manufacturer/tradename

Millipore

growth mode

N/A

technique(s)

cell analysis: suitable

shipped in

liquid nitrogen

storage temp.

−196°C

Application

Each vial contains ≥1X10⁶ viable cells.Cells are tested negative for infectious diseases by a Rat/Mouse Essential CLEAR Panel by Charles River Animal Diagnostic Services.Cells are verified to be of rat origin and negative for inter-species contamination from mouse, human, Chinese hamster, Golden Syrian hamster, and Non-human Primate (NHP) as assessed by a Contamination Clear panel by Charles River Animal Diagnostic ServicesCells are negative for mycoplasma contamination.
Glioma is one of the most common brain tumors and is highly aggressive and resistant to standard treatments. Immunotherapy is one of the more promising treatment options since the discovery of lymphatics in the central nervous system allows for penetration of the blood-brain barrier (1). Cellular models that both recapitulate in vivo characteristics of gliomas and are amenable to immunotherapeutic modalities are thus of great value in advancing understanding of gliomas and their potential treatment.The CNS-1 rat glioma cell line is a well-established model that closely mimics the invasive behavior and growth characteristics of human gliomas (2). CNS-1 cells are derived from an N-nitroso-N-methylurea-induced intracranial tumor of a male Lewis rat and are histocompatible with this inbred rat strain, facilitating a range of in vivo studies. Tumors derived from injection of CNS-1 cells display diffuse infiltrative patterns similar to human glioblastoma multiforme (2,3). CNS-1 cells are positive for glial fibrillary acid protein (GFAP), retinoic acid receptor α, and neural cell adhesion molecule NCAM-140 (2,4). CNS-1 cells exhibit multiple chromosome loss and rearrangements reflecting genetic instability common in many human gliomas (2). CNS-1 rat glioma cells are a highly relevant model with wide applications for glioma immunotherapies.CNS-1 cell line originated from an N-nitroso-N-methylurea-induced intracranial tumor of a male Lewis rat (1). 1. Cancer Lett 2020, 476: 1-12.2. J Neurooncol 1994, 22(3): 191-200.3. J Neurooncol 2009, 94(3): 299-312.4. Cancer Res 1998, 58(9): 2020-2028.

Features and Benefits

CNS-1 rat glioma cells are a highly relevant model with wide applications for glioma immunotherapies.​

Storage and Stability

Store in liquid nitrogen. The cells can be cultured for at least 10 passages after initial thawing without significantly affecting the cell marker expression and functionality.

Other Notes

This product is intended for sale and sold solely to academic institutions for internal academic research use per the terms of the “Academic Use Agreement” as detailed in the product documentation. For information regarding any other use, please contact licensing@emdmillipore.com.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service