Skip to Content
Merck
All Photos(1)

Key Documents

PX0045

Supelco

Mineral oil

white

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12181504
EC Index Number:
232-455-8
bp:
300-500 °C/1,013 hPa
vapor pressure:
<0.1 hPa ( 20 °C)
Pricing and availability is not currently available.

vapor pressure

<0.1 hPa ( 20 °C)

form

liquid

color

white
APHA: ≤20

refractive index

n20/D 1.467 (lit.)

bp

300-500 °C/1,013 hPa

transition temp

flash point 200 °C

density

0.86 g/cm3 at 20 °C

shipped in

ambient

storage temp.

room temp

InChI key

AEOVEGJBKQQFOP-DDVLFWKVSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Saybolt viscosity 340-365 at 100F
white

Analysis Note

Color (APHA)20 maxDescriptionTo pass testFormLiquidInfrared SpectrumConforms to standardLimit of polynuclear compoundsTo pass testNeutralityTo pass testReadily carbonizable substancesTo pass testSolid paraffinTo pass testSolubilityTo pass testSpecific Gravity at 25/25°C0.864-0.881

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

455.0 °F

Flash Point(C)

235 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service