Skip to Content
Merck
All Photos(1)

Key Documents

HTS132RTA

Sigma-Aldrich

Ready-to-Assay 5-HT 2C Serotonin Family Receptor Frozen Cells

Human

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
eCl@ss:
32011203
NACRES:
NA.76
biological source:
human
manufacturer/tradename:
Ready-to-Assay

Product Name

Ready-to-Assay 5-HT 2C Serotonin Family Receptor Frozen Cells,

biological source

human

Quality Level

manufacturer/tradename

Ready-to-Assay

technique(s)

calcium flux assay: suitable

NCBI accession no.

detection method

fluorometric

General description

Millipore′s Ready-to-Assay GPCR frozen cells are designed for simple, rapid calcium assays with no requirement for intensive cell culturing. Millipore has optimized the freezing conditions to provide cells with high viability and functionality post-thaw. The user simply thaws the cells and resuspends them in media, dispenses cell suspension into assay plates and, following over night recovery, assays for calcium response.

5-Hydroxytryptamine (5-HT, also commonly known as serotonin) is synthesized in enterochromaffin cells in the intestine and in serotonergic nerve terminals. In the periphery, 5-HT mediates gastrointestinal motility, platelet aggregation, and contraction of blood vessels. Many functions of the central nervous system are influenced by 5-HT, including sleep, motor activity, sensory perception, arousal and appetite. A family of 12 GPCRs and one ion channel mediate the biological effects of 5-HT (Hoyer et al., 1994). 5-HT2C, which couples to Gq in most cells to stimulate intracellular calcium, is prominently expressed in brain and appears to modulate depression, anxiety and appetite (Miller, 2005; Serretti et al., 2004; Wood, 2003). The mRNA encoding 5-HT2C undergoes selective RNA editing that changes 4 amino acids in the second intracellular loop; these changes result in alteration of efficiency of coupling to G proteins. Alterations in editing of 5-HT2C have been detected in victims of suicidal depression and in mice treated with the SSRI, fluoxetine (Tohda et al., 2006). Millipore′s cloned human 5-HT2C -expressing cell line is made in the Chem-1 host, which supports high levels of recombinant 5-HT2C expression on the cell surface for functional detection via the calcium signaling pathway. Thus, the cell line is an ideal tool for screening for agonists, antagonists and modulators at 5-HT2C.

Cell Line Description

  • GPCR Cell Lines
  • Host cells: Chem-1

Application

Human 5-HT2C GPCR frozen cells for Calcium Flux FLIPR Assays.

Biochem/physiol Actions

  • GPCR Class: A
  • Protein Target: 5-HT2C
  • Target Sub-Family: Serotonin

Components

Pack contains 2 vials of mycoplasma-free cells, 1 ml per vial.
Fifty (50) mL of Media Component.

Disclaimer


This product contains genetically modified organisms (GMO).
Within the EU GMOs are regulated by Directives 2001/18/EC and 2009/41/EC of the European Parliament and of the Council and their national implementation in the member States respectively.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service