Skip to Content
Merck
All Photos(2)

Documents

EZFREAG57

Millipore

EZ-Fluo Reagent Kit

For fluorescence-based microbial detection technology

Synonym(s):

Fluorescence-basedstaining kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116130
eCl@ss:
32160606
NACRES:
NA.85

Quality Level

manufacturer/tradename

EZ-Fluo

packaging

pkg of -57 tests

storage condition

room temperature

technique(s)

Rapid Sterility Testing: suitable

input

cell

application(s)

food and beverages

detection method

fluorometric

shipped in

ambient

storage temp.

15-25°C

General description

EZ-Fluo Reagent Kit is a part of the EZ-Fluo Rapid Detection System to address contamination events sooner. The kit consists of a reconstitution reagent, fluorescence reagent, staining buffer, plastic storage bag, and transfer pipettes to operate 57 tests.The EZ-Fluo method is based on a unique, non-destructive, fluorescent marker. After filtration and incubation of the recovered microbes, the staining reagent is applied. Initially, the reagent is colorless but on interaction with viable and culturable cells, a ubiquitous intracellular enzyme produced by the living cells cleaves the substrate and releases the fluorophore. On accumulation of the fluorophore, the signal is amplified and can be detected with the EZ-Fluo reader.

Application

The EZ-Fluo Reagent Kit, used as part of the EZ-Fluoez-fluo Rapid Detection System offers an efficient method to detect microbial contamination in filtrable samples such as raw materials,In-process samples, final products, and environmental samples.

Features and Benefits

  • Rapid and accurate quantitative detection of contaminants
  • The staining procedure is non-destructive
  • A reliable alternative for the detection of microbial contaminants

Legal Information

EZ-FLUO is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service