Skip to Content
Merck
All Photos(1)

Key Documents

8.18378

Sigma-Aldrich

(S)-(-)-ß-Citronellol

for synthesis

Synonym(s):

(S)-(-)-ß-Citronellol, (S)-(-)-3,7-Dimethyl-6-octen-1-ol, ß-Rhodinol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H20O
CAS Number:
Molecular Weight:
156.27
MDL number:
UNSPSC Code:
12352104
EC Index Number:
231-415-7
NACRES:
NA.22

Quality Level

Assay

≥98.0% (GC)

form

liquid

autoignition temp.

240 °C

potency

3450 mg/kg LD50, oral (Rat)
2650 mg/kg LD50, skin (Rabbit)

bp

109-111 °C/13 hPa

transition temp

flash point 101 °C (DIN 51758)

density

0.855 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

OCC[C@H](CCC=C(C)C)C

InChI

1S/C10H20O/c1-9(2)5-4-6-10(3)7-8-11/h5,10-11H,4,6-8H2,1-3H3/t10-/m0/s1

InChI key

QMVPMAAFGQKVCJ-JTQLQIEISA-N

Application

  • Influence of foliar fertilizers application on the volatile composition of red wines: This research discusses how foliar fertilization impacts the levels of ß-citronellol in red wines, noting that enhanced foliar nutrition can lead to increased concentrations of this compound. The study also examines the role of yeast metabolism in alcoholic fermentation, providing insights beneficial to both academia and industry focused on agricultural practices and fermentation science (D Dimitrov et al., 2023).

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.854 - 0.856
Spec. rotation [α²0/D (undiluted): -5.5 - -3.5 °
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

213.8 °F - DIN 51758

Flash Point(C)

101 °C - DIN 51758


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service