573126
STAT3 Inhibitor XI, STX-0119
The STAT3 Inhibitor XI, STX-0119 controls the biological activity of STAT3. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.
Synonym(s):
STAT3 Inhibitor XI, STX-0119, N-(5-(Furan-2-yl)-1,3,4-oxadiazol-2-yl)-2-phenylquinoline-4-carboxamide
About This Item
Recommended Products
Quality Level
Assay
≥90% (HPLC)
form
solid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
protect from light
color
gray to off-white
solubility
DMSO: soluble
shipped in
ambient
storage temp.
2-8°C
SMILES string
O=C(NC1=NN=C(C2=CC=CO2)O1)C3=CC(C4=CC=CC=C4)=NC5=C3C=CC=C5
InChI key
MNPXTRXFUMGQLK-UHFFFAOYSA-N
General description
Packaging
Warning
Other Notes
Matsuno, K., et al. 2010. ACS Med. Chem. Lett.1, 371.
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service