Skip to Content
Merck
All Photos(2)

Key Documents

1.07711

Supelco

Sea sand

extra pure

Synonym(s):

Sea sand

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
O2Si
CAS Number:
Molecular Weight:
60.08
MDL number:
UNSPSC Code:
11111701
EC Index Number:
238-878-4
NACRES:
NB.51
Pricing and availability is not currently available.

Quality Level

form

solid

impurities

≤0.5% HCl solubles

loss

≤0.5% loss on ignition, 900 °C

particle size

0.1-0.315 mm (>=90%)

pH

5-8 (20 °C, 400 g/L in H2O, slurry)

bp

>2200 °C/1013 hPa

mp

1713 °C

density

2.65 g/cm3 at 20 °C

bulk density

1400 kg/m3

anion traces

chloride (Cl-): ≤0.01%

storage temp.

no temp limit

SMILES string

[Si](=O)=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
1.07712SX0070SX0075
Sea sand extra pure

1.07711

Sea sand

Sea sand purified by acid and calcined for analysis

1.07712

Sea sand

Sand Standard

SX0070

Sand

Sand Standard, washed and dried

SX0075

Sand

form

solid

form

solid

form

granular

form

granular

pH

5-8 (20 °C, 400 g/L in H2O, slurry)

pH

5-8 (20 °C, 400 g/L in H2O, slurry)

pH

5-8 (20 °C, 400 g/L in H2O)

pH

5-8 (20 °C, 400 g/L in H2O)

Quality Level

200

Quality Level

200

Quality Level

-

Quality Level

-

impurities

≤0.5% HCl solubles

impurities

≤0.2% HCl solubles

impurities

-

impurities

≤0.2% HCl solubles, ≤0.2% volatiles

loss

≤0.5% loss on ignition, 900 °C

loss

≤0.05% loss on ignition, 900 °C

loss

-

loss

-

Analysis Note

Substances soluble in hydrochloric acid: ≤ 0.5 %
Chloride (Cl): ≤ 0.01 %
Loss on ignition (900 °C): ≤ 0.5 %
Particle size (0.1 - 0.315 mm): ≥ 90 %

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service