1.04326
Tryptic Soy Broth - Dehydrated Culture Media
NutriSelect® Prime, EP, USP, JP, ISO, FDA BAM, powder, Suitable for use in Halal and Kosher certified processes
Synonym(s):
NutriSelect® prime Tryptic Soy Broth halal acc. EP, USP, JP, ISO and FDA-BAM, TSB, soy broth, CASO Broth, Casein-peptone Soymeal-peptone Broth, tryptone soya broth, CASO Broth, Casein Soya Broth, Soybean Casein digest Broth, TSB, Tryptone Soya Broth
About This Item
Agency
EP
ISO
JP
USP
Quality Level
reg. compliance
FDA BAM
form
powder
manufacturer/tradename
NutriSelect® Prime
technique(s)
microbiological culture: suitable
pH
7.3 ± 0.2 (25.0 °C)
application(s)
pharmaceutical
storage temp.
15-25°C
suitability
nonselective for
Looking for similar products? Visit Product Comparison Guide
General description
Application
Legal Information
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Skin Sens. 1
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service