Key Documents
Z285889
Sigma-Aldrich
E-Z Press
About This Item
UNSPSC Code:
12000000
Pricing and availability is not currently available.
L × D × H
7 1/2 in. × 6 in. × 15 1/2 in.
plate diam.
4 in.
weight
49.9 lb
General description
• Built-in safety shield
• Ergonomic design
• Light weight
12 ton laboratory press with unibody construction designed to work with the Aldrich macro-micro and similar KBr pellet dies. Built-in pressure gauge and pressure release valve provide accurate control of pressure. Press includes removable handle and base.
Legal Information
E-Z Press is a trademark of International Crystal Laboratories Corp.
Choose from one of the most recent versions:
Certificates of Analysis (COA)