Documents
Z275271
Sigma-Aldrich
Cannon-Fenske viscometer tube
size 50
About This Item
UNSPSC Code:
41111700
NACRES:
PB.36
size
50
description
constant ~ 0.004 cSt/s
manufacturer/tradename
Ace Glass 798835
viscosity measuring range
0.8-4 cSt
General description
Obtain kinematic viscosities using tubes designed for ASTM Method D445 and D2515. Viscosities can be measured with a precision of ±0.2% quickly and easily.
Requires approximately 7 mL of sample. Certificate of Calibration supplied.