WHA2103
Whatman® PolyVENT and Sterivent intregal filters
Sterivent 36, inlet 3/8 to 1/2 inch, outlet Stepped Hosebarb Outlet
Synonym(s):
Z741345
About This Item
Recommended Products
material
PTFE membrane
polypropylene housing
sterility
non-sterile
manufacturer/tradename
Whatman 2103
Whatman Article No. 28418681 (US reference)
packaging
1 ea of
diam.
36 mm
pore size
0.2 μm
fitting
inlet 3/8 to 1/2 inch
outlet Stepped Hosebarb Outlet
Looking for similar products? Visit Product Comparison Guide
Related Categories
1 of 4
This Item | 1.09441 | 1.09439 | 1.09435 |
---|---|---|---|
Certipur® | Certipur® | Certipur® | Certipur® |
certified reference material | certified reference material | certified reference material | certified reference material |
environmental | environmental | environmental | environmental |
1.0 (25 °C in H2O) | 1.0 (25 °C in H2O) | 7.0 (20 °C in H2O) | 4.0 (20 °C in H2O) |
1.00 g/cm3 at 20 °C | 1.00 g/cm3 at 20 °C | 1.01 g/cm3 at 20 °C | 1.01 g/cm3 at 20 °C |
General description
Whatman PolyVENT venting filter
Draining or filling of incubators, fermentation tanks, and other vessels requires a venting filter capable of preventing bacterial contamination. With an integral PTFE filter membrane, Whatman PolyVENT acts as an industrial air filter media for sterilization* of gases entering bioreactors such as fermentation tanks.
Features and Benefits
- 0.2 µm hydrophobic PTFE air filters are ideal industrial air filter media
- Validated for 50 steam autoclave cycles; compatible with ethylene oxide (EtO)
- Testable by water break through (WBT) test or bubble point testing
- Passes USP Class VI biosafety tests for plastics
- Manufactured in clean room facilities
- Filtration area 500, 1000, and 2000 cm² to support a range of venting applications
Other Notes
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service