Skip to Content
Merck
All Photos(1)

Key Documents

W372218

Sigma-Aldrich

cis-5-Octen-1-ol

≥97%, stabilized, FG

Synonym(s):

(Z)-Oct-5-en-1-ol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5CH=CH(CH2)4OH
CAS Number:
Molecular Weight:
128.21
FEMA Number:
3722
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
2.113
NACRES:
NA.21
Organoleptic:
fresh; green; melon
grade:
FG
Fragrance grade
Halal
Kosher
biological source:
synthetic
Agency:
follows IFRA guidelines
meets purity specifications of JECFA
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 872/2012

Assay

≥97%

contains

0.01-0.20% α-tocopherol as stabilizer (synthetic)

refractive index

n20/D 1.448 (lit.)

bp

95 °C/25 mmHg (lit.)

density

0.849 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

fresh; green; melon

SMILES string

[H]\C(CC)=C(/[H])CCCCO

InChI

1S/C8H16O/c1-2-3-4-5-6-7-8-9/h3-4,9H,2,5-8H2,1H3/b4-3-

InChI key

VDHRTASWKDTLER-ARJAWSKDSA-N

Related Categories

General description

cis-5-Octen-1-ol has been identified as one of the volatile aroma components of apples. It may also be a potent attractant to fruit fly.

Application


  • RIFM fragrance ingredient safety assessment, cis-5-octen-1-ol CAS Registry Number 64275-73-6.: This comprehensive safety assessment of cis-5-octen-1-ol evaluates its use in fragrances, ensuring its safety for consumers. The study highlights the compound′s potential applications in the fragrance industry, emphasizing its non-toxic nature and suitability for various consumer products (Api et al., 2018).


Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

190.4 °F - closed cup

Flash Point(C)

88 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Quantitative metabolic profiling of grape, apple and raspberry volatile compounds (VOCs) using a GC/MS/MS method
Vrhovsek U, et al.
Journal of Chromatography. B, Biomedical Applications, 966, 132-139 (2014)
Synthesis, spectra, odor properties, and structural relationship of (Z)-6-Nonenal and (Z)-5-octen-1-ol to fruit fly attractants.
Seifert RM
Journal of Agricultural and Food Chemistry, 29(3), 647-649 (1981)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service