Skip to Content
Merck
All Photos(3)

Key Documents

M29959

Sigma-Aldrich

4-Methylbenzophenone

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4COC6H5
CAS Number:
Molecular Weight:
196.24
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

Assay

99%

bp

326 °C (lit.)

mp

56.5-57 °C (lit.)

SMILES string

Cc1ccc(cc1)C(=O)c2ccccc2

InChI

1S/C14H12O/c1-11-7-9-13(10-8-11)14(15)12-5-3-2-4-6-12/h2-10H,1H3

InChI key

WXPWZZHELZEVPO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Growth of 4-methylbenzophenone (4MB) single crystals by Czochralski method and its structural, mechanical and optical characterization: This study discusses the successful growth of 4-methylbenzophenone crystals and their properties, advancing materials science applications (Ramachandran et al., 2018).
  • Synthesis and characteristics of photopolymerized benzophenone: The synthesis of 4-methylbenzophenone acrylate for low migration applications in polymers was explored, suggesting its utility in safer material design (Tang et al., 2017).

Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2 - STOT RE 2 Oral

Target Organs

Kidney,Liver

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stanislav Gobec et al.
Bioorganic & medicinal chemistry letters, 15(23), 5170-5175 (2005-09-27)
Nonsteroidal anti-inflammatory drugs (NSAIDs) like indomethacin, flufenamic acid, and related compounds have been recently identified as potent inhibitors of AKR1C3. We report that some other NSAIDs (diclofenac and naproxen) also inhibit AKR1C3, with the IC(50) values in the low micromolar

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service