Skip to Content
Merck
All Photos(1)

Key Documents

GF50131651

Polyimide

(PI), tube, outside diameter 3.0mm, L 1000 mm, wall thickness 0.03 mm

Synonym(s):

IM307300

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352100
NACRES:
NA.23

Pricing and availability is not currently available.

form

tubes

manufacturer/tradename

Goodfellow 501-316-51

L

1000 mm

wall thickness

0.03 mm

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
GF85451126GF77133090GF57893675
manufacturer/tradename

Goodfellow 501-316-51

manufacturer/tradename

Goodfellow 854-511-26

manufacturer/tradename

Goodfellow 771-330-90

manufacturer/tradename

Goodfellow 578-936-75

form

tubes

form

tubes

form

tubes

form

tubes

wall thickness

0.03 mm

wall thickness

0.05 mm

wall thickness

0.025 mm

wall thickness

0.075 mm

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service