DWKCT1000P
Azlon Hexagonal Base PMP Cylinders with Molded Graduations
class B, capacity 1000 mL , with +/10mL tolerance
Synonym(s):
CT1000P
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121805
NACRES:
NA.54
Recommended Products
feature
class B
graduated
packaging
pkg of 1 ea
manufacturer/tradename
Azlon CT1000P
capacity
1000 mL , with +/10mL tolerance
Looking for similar products? Visit Product Comparison Guide
Application
Suitable for the measurement and dispensing of liquids in the laboratory
Features and Benefits
Conform to ISO 6706
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service