CLS982513
Pyrex® culture tubes with screw-cap
O.D. × L 13 mm × 100 mm
Synonym(s):
Corning® 9825
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121706
NACRES:
NB.43
material
borosilicate glass
cap (Rubber Liner)
round bottom tube
thread size
13-415
sterility
non-sterile
packaging
pack of 144
case of 288
manufacturer/tradename
Corning 9825-13
O.D. × L
13 mm × 100 mm
color
clear
suitability
suitable for (culture)
Looking for similar products? Visit Product Comparison Guide
General description
These Pyrex culture tubes are supplied with deep-form 18-415 GPI threaded phenolic caps to facilitate handling and sealing after autoclaving. Caps are fabricated from a special formula resistant to the effects of high temperatures and steam in autoclaving and are provided with a white rubber liner. The white enamel marking spot gives an excellent surface for pencil notations.
Legal Information
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service