Skip to Content
Merck
All Photos(1)

Key Documents

907308

Sigma-Aldrich

(3-(1,1-Difluoro-3-(triisopropylsilyl)prop-2-yn-1-yl)-3H-diazirin-3-yl)methanol

≥95%

Synonym(s):

Hydroxyl diazirine difluoro TIPS-alkyne, MI-PAL, Minimally interfering photoaffinity label, Photo-click tag, Probe building block

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H24F2N2OSi
CAS Number:
Molecular Weight:
302.44
MDL number:
UNSPSC Code:
12352200
Pricing and availability is not currently available.

Assay

≥95%

form

liquid

availability

available only in USA

storage temp.

−20°C

SMILES string

[Si](C(C)C)(C(C)C)(C(C)C)C#CC(F)(F)C1(N=N1)CO

InChI

1S/C14H24F2N2OSi/c1-10(2)20(11(3)4,12(5)6)8-7-13(15,16)14(9-19)17-18-14/h10-12,19H,9H2,1-6H3

InChI key

SKAPASYQQQYNJK-UHFFFAOYSA-N

Application

(3-(1,1-Difluoro-3-(triisopropylsilyl)prop-2-yn-1-yl)-3H-diazirin-3-yl)methanol is a minimally interfering photo-affinity label (MI-PAL). Photoaffinity labeling is of growing interest in chemical biology and drug discovery research to characterize the non-covalent interactome and is used in protein profiling; the study of protein-protein interactions and complexes; and target engagement, identification, or validation studies. This MI-PAL contains a diazirine for UV light-induced (∼365 nm) covalent modification of a biological target with potential for downstream applications via the click chemistry-enabling alkyne tag. Its small size exhibits minimal perturbation of the interactions between proteins and the ligand or probe. The Woo lab demonstrated the electronic tuning of this MI-PAL with the placement of stability-imparting fluorine groups adjacent to the diazirine and alkyne. The alkyl fluorine ion losses are also a useful marker in collision-induced dissociation (CID) mass spectrometry experiments. Use alone or in parallel with other multi-functional building blocks to discover the optimal probe for your chemical biology experiments.

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Self-react. C

Storage Class Code

5.2 - Organic peroxides and self-reacting hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service