Skip to Content
Merck
All Photos(2)

Key Documents

906115

Sigma-Aldrich

2,7-Dibromoacridone

Synonym(s):

2,7-Dibromoacridin-9(10H)-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H7Br2NO
CAS Number:
Molecular Weight:
353.01
MDL number:
UNSPSC Code:
12352101
NACRES:
NA.22
Pricing and availability is not currently available.

form

powder

reaction suitability

reagent type: catalyst
reaction type: Photocatalysis

SMILES string

Brc1cc2c(nc3c(c2O)cc(cc3)Br)cc1

InChI

1S/C13H7Br2NO/c14-7-1-3-11-9(5-7)13(17)10-6-8(15)2-4-12(10)16-11/h1-6H,(H,16,17)

InChI key

ALEGIHZSVGKVFN-UHFFFAOYSA-N

Application

2,7-Dibromoacridone is used in the synthesis of series of acridinium photocatalysts.
Product can be used with our line of photoreactors: Including Penn PhD (Z744035) & SynLED 2.0 (Z744080)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Norihiro Mizoshita et al.
Chemical communications (Cambridge, England), 46(48), 9235-9237 (2010-11-03)
Polycondensation of 9-mesityl-10-methylacridinium-bridged organosilane in the presence of a nonionic surfactant yielded a mesostructured organosilica solid with a functional framework that exhibited long-lived photoinduced charge separation.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service